missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Phospho-Nbs1 (S343) Antibody, R&D Systems™

Rabbit Polyclonal Antibody
Brand: R&D Systems AF4944-SP
This item is not returnable.
View return policy
Description
Nbs1 Polyclonal specifically detects Nbs1 in Human samples. It is validated for Western Blot.
Specifications
| Nbs1 (p ser343) | |
| Polyclonal | |
| Western Blot 1 ug/mL | |
| ATV, AT-V1, AT-V2, Cell cycle regulatory protein p95, FLJ10155, MGC87362, NBN, NBS, NBS1P95, nibrin, Nijmegen breakage syndrome 1 (nibrin), Nijmegen breakage syndrome protein 1, p95, p95 protein of the MRE11/RAD50 complex | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| 4683 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| Western Blot | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS and Sodium Azide with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with Sodium Azide | |
| NBN | |
| Phosphopeptide containing human Nbs1 S343 site | |
| 25 μg | |
| Primary | |
| Detects human Nbs1 when phosphorylated at S343. Does not recognize Nbs1 when unphosphorylated at S343. | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction