Learn More
Human Phospho-NFATC1 (S172) Antibody, R&D Systems™
Mouse Monoclonal Antibody
Brand: R&D Systems MAB5640-SP
This item is not returnable.
View return policy
Description
NFATC1/NFAT2 Monoclonal specifically detects NFATC1/NFAT2 in Human samples. It is validated for Western Blot.
Specifications
| NFATC1/NFAT2 (p ser172) | |
| Monoclonal | |
| LYOPH | |
| Western Blot 1 ug/mL | |
| MGC138448, NFAT transcription complex cytosolic component, NFAT2, NFAT2cytoplasmic 1, NFATC, NF-ATC, NFATc1, NF-ATc1, nuclear factor of activated T-cells, cytoplasmic, calcineurin-dependent 1 | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 4772 | |
| Sterile PBS to a final concentration of 0.5 mg/mL. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG1 |
| Western Blot | |
| 679340 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| NFATC1 | |
| Phosphopeptide containing the human S172 site. | |
| 25 μg | |
| Primary | |
| Detects human NFATC1 in direct ELISAs. | |
| Human | |
| Purified |