missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Phospho-SMC1 (S966) Antibody, R&D Systems™

Rabbit Polyclonal Antibody
Brand: R&D Systems AF2677-SP
This item is not returnable.
View return policy
Description
SMC1 Polyclonal specifically detects SMC1 in Human samples. It is validated for Western Blot.
Specifications
| SMC1 (p ser966) | |
| Polyclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| SMC1A | |
| Phosphopeptide containing human SMC1 S966 site | |
| 25 μg | |
| Primary | |
| Detects human SMC1 when phosphorylated at S966. | |
| Human | |
| IgG |
| Western Blot | |
| LYOPH | |
| Western Blot 0.5 ug/mL | |
| CDLS2, DXS423ESMC1 structural maintenance of chromosomes 1-like 1 (yeast), KIAA0178DKFZp686L19178, SB1.8, SB1.8MGC138332, segregation of mitotic chromosomes 1, SMC protein 1A, SMC1 (structural maintenance of chromosomes 1, yeast)-like 1, SMC1 structural maintenance of chromosomes 1-like 1, SMC1A, SMC-1A, SMC-1-alpha, SMC1L1, SMC1SMC1alpha, SMCB, structural maintenance of chromosomes 1A, structural maintenance of chromosomes protein 1A | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| 8243 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction