missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Phospho-STAT2 (Y689) Antibody, R&D Systems™

Rabbit Monoclonal Antibody
Brand: R&D Systems MAB2890-SP
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
STAT2 Monoclonal specifically detects STAT2 in Human samples. It is validated for Western Blot, Simple Western, Flow Cytometry.
Spécification
| STAT2 (p Tyr689) | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with Sodium Azide | |
| STAT2 | |
| Phosphopeptide containing the human STAT2 Y689 site | |
| 25 μg | |
| Primary | |
| Detects human phospho-STAT2 (Y689) in direct ELISAs and Western blots. | |
| Human | |
| Purified |
| Western Blot, Flow Cytometry | |
| 1021D | |
| Western Blot 0.05 ug/mL, Simple Western 0.5 ug/mL, Flow Cytometry 0.25 ug/mL | |
| interferon alpha induced transcriptional activator, ISGF-3, MGC59816, p113, signal transducer and activator of transcription 2, signal transducer and activator of transcription 2, 113kD, signal transducer and activator of transcription 2, 113kDa, STAT113 | |
| Rabbit | |
| Protein A or G purified from cell culture supernatant | |
| RUO | |
| 6773 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG |
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
For Research Use Only
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu