missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Phospho-STAT3 (S727) Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems MAB4934
This item is not returnable.
View return policy
Description
STAT3 Monoclonal specifically detects STAT3 in Human samples. It is validated for Western Blot, Intracellular Staining by Flow Cytometry.
Specifications
| STAT3 (p ser727) | |
| Monoclonal | |
| LYOPH | |
| Western Blot 1 ug/mL, Intracellular Staining by Flow Cytometry 0.25 ug/10^6 cells | |
| Acute-phase response factor, APRFMGC16063, DNA-binding protein APRF, FLJ20882, HIES, signal transducer and activator of transcription 3, signal transducer and activator of transcription 3 (acute-phase responsefactor) | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 6774 | |
| Sterile PBS to a final concentration of 0.5 mg/mL. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2b |
| Western Blot, Flow Cytometry | |
| 788335 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| STAT3 | |
| Phosphopeptide containing the human STAT3 S727 site | |
| 100 μg | |
| Primary | |
| Detects human STAT3 when phosphorylated at S727. | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction