Learn More
Human Phospho-STAT3 (Y705) Antibody, R&D Systems™
Rat Monoclonal Antibody has been used in 2 publications
Brand:  R&D Systems MAB4607
Product Code. 18101811
  • ONLINE EXCLUSIVE PRICE
    € 494.00
    € 467.25 / 100µg
    Save € 26.75
    5% Off
Description
STAT3 Monoclonal specifically detects STAT3 in Human samples. It is validated for Western Blot.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
STAT3 (p tyr705)
Monoclonal
LYOPH
Western Blot 2 ug/mL
Acute-phase response factor, APRFMGC16063, DNA-binding protein APRF, FLJ20882, HIES, signal transducer and activator of transcription 3, signal transducer and activator of transcription 3 (acute-phase responsefactor)
Rat
Protein A or G purified from hybridoma culture supernatant
RUO
6774
Sterile PBS to a final concentration of 0.5 mg/mL.
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
IgG2b
Western Blot
805435
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
STAT3
Phosphopeptide containing the human STAT3 Y705 site
100 μg
Primary
Detects human STAT3 when phosphorylated at Y705.