missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human PI 3-Kinase p110 delta Antibody, R&D Systems™

Mouse Monoclonal Antibody has been used in 2 publications
Brand: R&D Systems MAB2687-SP
This item is not returnable.
View return policy
Description
PI 3-Kinase p110 delta Monoclonal specifically detects PI 3-Kinase p110 delta in Human samples. It is validated for Western Blot.
Specifications
| PI 3-Kinase p110 delta | |
| Monoclonal | |
| LYOPH | |
| Western Blot 1 ug/mL | |
| O00329 | |
| PIK3CD | |
| E. coli-derived recombinant human PI 3-Kinase p110 delta Cys391-Phe542 Accession # O00329 | |
| 25 μg | |
| Primary | |
| Detects endogenous human PI 3-kinase p110 delta in Western blots. | |
| Human | |
| Purified |
| Western Blot | |
| 275144 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| EC 2.7.1, EC 2.7.1.153, p110D, P110DELTA, Phosphatidylinositol-4,5-bisphosphate 3-kinase 110 kDa catalytic subunit delta, phosphatidylinositol-4,5-bisphosphate 3-kinase catalytic subunit delta isoform, phosphoinositide-3-kinase C, phosphoinositide-3-kinase, catalytic, delta polypeptide, PI 3Kinase p110 delta, PI3K, PI3K-delta, PI3-kinase p110 subunit delta, PI3-kinase subunit delta, PIK3CD, PtdIns-3-kinase subunit delta, PtdIns-3-kinase subunit p110-delta | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 5293 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction