missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Plexin B1 PE-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems FAB37491P
This item is not returnable.
View return policy
Description
Plexin B1 Monoclonal specifically detects Plexin B1 in Human samples. It is validated for Flow Cytometry.
Specifications
| Plexin B1 | |
| Monoclonal | |
| R-PE | |
| Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide | |
| plexin B1, PLXN5, PLXNB1, SEP, Transmembrane Protein SEP | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 5364 | |
| Human | |
| Purified |
| Flow Cytometry | |
| 559830 | |
| Flow Cytometry 10 uL/10^6 cells | |
| O43157 | |
| PLXNB1 | |
| NS0 mouse myeloma cell line transfected with human Plexin B1 Accession # O43157 | |
| 100 Tests | |
| Primary | |
| Detects human Plexin B1 in direct ELISAs. | |
| Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction