missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Plexin B2 Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems MAB53292
This item is not returnable.
View return policy
Description
Plexin B2 Monoclonal specifically detects Plexin B2 in Human samples. It is validated for Western Blot.
Specifications
| Plexin B2 | |
| Monoclonal | |
| LYOPH | |
| Western Blot 2 ug/mL | |
| O15031 | |
| PLXNB2 | |
| Chinese hamster ovary cell line CHO-derived recombinant human Plexin B2 Leu20-Arg1160 Accession # O15031 | |
| 100 μg | |
| Primary | |
| Detects human Plexin B2 in direct ELISAs. In direct ELISAs, no cross-reactivity with recombinant mouse Plexin B2 or recombinant human Plexin B3 is observed. | |
| Human | |
| Purified |
| Western Blot | |
| 537219 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| MM1, MM1KIAA0315dJ402G11.3, Nbla00445, PLEXB2, plexin B2, plexin-B2, PLXNB2 | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 23654 | |
| Sterile PBS to a final concentration of 0.5 mg/mL. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction