missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Plexin D1 Alexa Fluor 488-conjugated Antibody, R&D Systems™

Goat Polyclonal Antibody
Brand: R&D Systems FAB4160G
This item is not returnable.
View return policy
Description
Plexin D1 Polyclonal specifically detects Plexin D1 in Human samples. It is validated for Flow Cytometry.
Specifications
| Plexin D1 | |
| Polyclonal | |
| Flow Cytometry 5 uL/10^6 cells | |
| KIAA0620MGC75353, PLEXD1, plexin D1, plexin-D1, PLXND1 | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 23129 | |
| Human | |
| IgG |
| Flow Cytometry | |
| Alexa Fluor 488 | |
| Q9Y4D7 | |
| PLXND1 | |
| Mouse myeloma cell line NS0-derived recombinant human Plexin D1 Leu47-Ala1271 Accession # Q9Y4D7 | |
| 100 Tests | |
| Primary | |
| Detects human Plexin D1 in direct ELISAs and Western blots. In direct ELISAs and Western blots, less than 1% cross-reactivity with recombinant mouse (rm) Plexin A1, rmPlexin A3, and recombinant human Plexin B1 is observed. | |
| Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction