missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human PMEL Antibody, R&D Systems™

Mouse Monoclonal Antibody
€ 248.00 - € 561.00
Specifications
| Antigen | PMEL17/SILV |
|---|---|
| Clone | 1067144 |
| Dilution | Immunohistochemistry 5-15 ug/mL |
| Applications | Immunohistochemistry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18190101
![]() |
R&D Systems
MAB11439-100 |
100 μg |
€ 561.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18169510
|
R&D Systems
MAB11439-SP |
25 μg |
€ 248.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Descripción
PMEL17/SILV Monoclonal antibody specifically detects PMEL17/SILV in Human samples. It is validated for ImmunohistochemistryEspecificaciones
| PMEL17/SILV | |
| Immunohistochemistry 5-15 ug/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied either lyophilized or as a 0.2 μm filtered solution in PBS. | |
| D12S53EP1, gp100, ME20, ME20M, ME20-M, melanocyte protein mel 17, Melanocyte protein Pmel 17, Melanocytes lineage-specific antigen GP100, Melanoma-associated ME20 antigen, melanosomal matrix protein17, PMEL17, PMEL17P100, premelanosome proteinME20M, SI, SIL, SILV, silver (mouse homolog) like, silver homolog (mouse), Silver locus protein homolog, silver, mouse, homolog of, SILVPmel17 | |
| Chinese Hamster Ovary cell line, CHO-derived human PMEL, Lys25-Gln595, Accession # P40967 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. |
| 1067144 | |
| Immunohistochemistry | |
| Unconjugated | |
| Mouse | |
| Human | |
| P40967 | |
| 6490 | |
| IgG2b | |
| Protein A or G purified from hybridoma culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20°C to -70°C as supplied. 1 month, 2°C to 8°C under sterile conditions after reconstitution. 6 months, -20°C to -70°C under sterile conditions after reconstitution. |
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto