missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human PP1 Inhibitor-2 Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems MAB4719
This item is not returnable.
View return policy
Description
PP1 Inhibitor-2 Monoclonal specifically detects PP1 Inhibitor-2 in Human samples. It is validated for Western Blot.
Specifications
| PP1 Inhibitor-2 | |
| Monoclonal | |
| LYOPH | |
| Western Blot 1 ug/mL | |
| P41236 | |
| PPP1R2 | |
| E. coli-derived recombinant human PP1 Inhibitor-2 Met1-Ser205 Accession # P41236 | |
| 100 μg | |
| Primary | |
| Detects human PP1 Inhibitor-2 in direct ELISAs and Western blots. | |
| Human | |
| Purified |
| Western Blot | |
| 488123 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| IPP2, IPP-2, IPP2phosphatase inhibitor 2, MGC87148, phosphoprotein phosphatase, PP1 Inhibitor2, PPP1R2, protein phosphatase 1, regulatory (inhibitor) subunit 2, protein phosphatase inhibitor 2 | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 5504 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction