missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human PP2C gamma Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems MAB5595-SP
This item is not returnable.
View return policy
Description
PP2C gamma/PPM1G Monoclonal specifically detects PP2C gamma/PPM1G in Human samples. It is validated for Western Blot, Immunohistochemistry.
Specifications
| PP2C gamma/PPM1G | |
| Monoclonal | |
| LYOPH | |
| Western Blot 0.25 ug/mL, Immunohistochemistry 8-25 ug/mL | |
| O15355 | |
| PPM1G | |
| E. coli-derived recombinant human PP2C gamma Met1-Asp546 Accession # O15355 | |
| 25 μg | |
| Primary | |
| Detects human PP2C gamma . | |
| Human | |
| Purified |
| Western Blot, Immunohistochemistry | |
| 528322 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| EC 3.1.3.16, MGC1675, MGC2870, PP2C, gamma, PP2CG, PP2Cgamma, PP2C-gamma, PPM1C, PPM1G, PPP2CG, Protein phosphatase 1C, protein phosphatase 1G, protein phosphatase 1G (formerly 2C), magnesium-dependent, gamma isoform, protein phosphatase 2, catalytic subunit, gamma isoform, protein phosphatase 2C gamma isoform, Protein phosphatase 2C isoform gamma, Protein phosphatase magnesium-dependent 1 gamma, protein phosphatase, Mg2+/Mn2+ dependent, 1G | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 5496 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction