missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human PPAP2C (aa 31-53) Control Fragment Recombinant Protein

Product Code. 30198671
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
This item is not returnable. View return policy

Product Code. 30198671

Brand: Invitrogen™ RP110221

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (100%), Rat (100%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-144872 (PA5-144872. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

Phosphatidic acid phosphatase type 2 (PAP2) was originally identified as a plasma membrane enzyme that catalyses the dephosphorylation of the putative second messenger, phosphatidic acid (PA) to diacylglycerol (DG). Subsequently, multiple isoforms of PAP2 were cloned [2-5]. It was found that these enzymes dephosphorylate a number of lipid phosphates in vitro other than PA, including the potent bioactive lipids, lysophosphatidic acid (LPA) and sphingosine 1-phosphate (S1P). Therefore, they have been renamed lipid phosphate phosphatases (LPPs). Currently, there are four members of this family called LPP1, LPP1a, LPP2 and LPP3.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number O43688
Concentration ≥5.0 mg/mL
For Use With (Application) Blocking Assay, Control
Formulation 1 M urea, PBS with no preservative; pH 7.4
Gene ID (Entrez) 8612
Name Human PPAP2C (aa 31-53) Control Fragment
Quantity 100 μL
Regulatory Status RUO
Gene Alias lipid phosphate phosphohydrolase 2; LPP2; LPR3; PAP2c; PAP-2 c; PAP2-G; PAP2-gamma; phosphatidate phosphohydrolase type 2 c; Phosphatidic acid phosphatase 2 c; phosphatidic acid phosphatase type 2 C; phosphatidic acid phosphohydrolase type 2 c; phospholipid phosphatase 2; PLPP2; Ppap2c; PRG2; PRG-2; type-2 phosphatidic acid phosphatase-gamma
Common Name PPAP2C
Gene Symbol PLPP2
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence PYKRGFYCGDDSIRYPYRPDTIT
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.