Learn More
R&D Systems™ Human PPAR gamma/NR1C3 (NP_056953) VersaClone cDNA
Click to view available options
Quantity:
10 μg
Förpackningsstorlek:
10µg
Beskrivning
The Human PPAR gamma/NR1C3 (NP_056953) VersaClone cDNA clone contains the complete ORF for PPAR gamma/NR1C3, along with a Kozak consensus sequence for optimal translation initiation. The gene insert is flanked with convenient multiple cloning sites which can be used to easily cut and transfer the gene cassette into your desired expression vector. The PPAR gamma/NR1C3 insert is sequence verified and the entire plasmid DNA and insert translated sequences are provided.
Specifikationer
Specifikationer
| Gene | hPPARG |
| Accession Number | NP_056953 |
| Concentration | 10 μg at 0.2 μg/μL |
| Formulation | cDNA is provided in 10 mM Tris-Cl |
| Insert Type | 1531bp |
| pH | 8.5 |
| Gene ID (Entrez) | 5468 |
| Content And Storage | Store the unopened product at -20°C to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date. |
| Species | Human |
| Quantity | 10 μg |
Korrigering av produktinnehåll
Din input är viktig för oss. Fyll i det här formuläret för att ge feedback relaterad till innehållet på denna produkt.
Produkttitel
Hittar du en möjlighet till förbättring?Dela en innehållskorrigering