missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human PPP1R13B (aa 720-866) Control Fragment Recombinant Protein

Product Code. 30195521
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
This item is not returnable. View return policy

Product Code. 30195521

Brand: Invitrogen™ RP88994

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (86%), Rat (86%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-52289 (PA5-52289. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

This gene encodes a member of the ASPP family of p53 interacting proteins. The protein contains four ankyrin repeats and an SH3 domain involved in protein-protein interactions. ASPP proteins are required for the induction of apoptosis by p53-family proteins. They promote DNA binding and transactivation of p53-family proteins on the promoters of proapoptotic genes. Expression of this gene is regulated by the E2F transcription factor.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q96KQ4
Concentration ≥5.0 mg/mL
For Use With (Application) Blocking Assay, Control
Formulation 1 M urea, PBS with no preservative; pH 7.4
Gene ID (Entrez) 23368
Name Human PPP1R13B (aa 720-866) Control Fragment
Quantity 100 μL
Regulatory Status RUO
Gene Alias AI449786; Apoptosis-stimulating of p53 protein 1; apoptosis-stimulating protein of p53, 1; ASPP1; AW545810; KIAA0771; p53BP2-like; p85; Ppp1r13b; Protein phosphatase 1 regulatory subunit 13 B; protein phosphatase 1, regulatory (inhibitor) subunit 13 B; protein phosphatase 1, regulatory subunit 13 B; Tp53bp2; transformation related protein 53 binding protein 2; Trp53bp2; tumor protein p53 binding protein, 2; tumor protein p53-binding protein, 2
Common Name PPP1R13B
Gene Symbol PPP1R13B
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence PNIQKLLYQRFNTLAGGMEGTPFYQPSPSQDFMGTLADVDNGNTNANGNLEELPPAQPTAPLPAEPAPSSDANDNELPSPEPEELICPQTTHQTAEPAEDNNNNVATVPTTEQIPSPVAEAPSPGEEQVPPAPLPPASHPPATSTNK
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.