missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human PPRC1 (aa 1263-1368) Control Fragment Recombinant Protein
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
Description
Highest antigen sequence indentity to the following orthologs: Mouse (85%), Rat (85%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-84352 (PA5-84352. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.
The protein encoded by this gene is similar to PPAR-gamma coactivator 1 (PPARGC1/PGC-1), a protein that can activate mitochondrial biogenesis in part through a direct interaction with nuclear respiratory factor 1 (NRF1). This protein has been shown to interact with NRF1. It is thought to be a functional relative of PPARGC1 that activates mitochondrial biogenesis through NRF1 in response to proliferative signals.
Specifications
Specifications
| Accession Number | Q5VV67 |
| Concentration | ≥5.0 mg/mL |
| For Use With (Application) | Blocking Assay, Control |
| Formulation | 1 M urea, PBS with no preservative; pH 7.4 |
| Gene ID (Entrez) | 23082 |
| Name | Human PPRC1 (aa 1263-1368) Control Fragment |
| Quantity | 100 μL |
| Regulatory Status | RUO |
| Gene Alias | A930018M19; BC013720; KIAA0595; mKIAA0595; peroxisome proliferative activated receptor, gamma, coactivator-related 1; peroxisome proliferator-activated receptor gamma coactivator-related protein 1; peroxisome proliferator-activated receptor gamma, coactivator-related 1; PGC-1-related coactivator; PPRC1; PRC; RP11-302K17.6 |
| Common Name | PPRC1 |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction