missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human PRDM15 (aa 1213-1344) Control Fragment Recombinant Protein
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
Description
Highest antigen sequence indentity to the following orthologs: Mouse (95%), Rat (95%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-55036 (PA5-55036. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.
The exact function of PRDM15 remains unknown.
Specifications
Specifications
| Accession Number | P57071 |
| Concentration | ≥5.0 mg/mL |
| For Use With (Application) | Blocking Assay, Control |
| Formulation | 1 M urea, PBS with no preservative; pH 7.4 |
| Gene ID (Entrez) | 63977 |
| Name | Human PRDM15 (aa 1213-1344) Control Fragment |
| Quantity | 100 μL |
| Regulatory Status | RUO |
| Gene Alias | C21orf83; E130018M06Rik; ORF62; PFM12; PFM15; PR domain 15; PR domain containing 15; PR domain zinc finger protein 15; PR domain-containing protein 15; PR/SET domain 15; PRDM15; Zfp298; Zinc finger protein 298; ZNF298 |
| Common Name | PRDM15 |
| Mehr anzeigen |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur