missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human/Primate GDF-15 Biotinylated Antibody, R&D Systems™
Goat Polyclonal Antibody has been used in 6 publications
Brand: R&D Systems BAF940
This item is not returnable.
View return policy
Description
GDF-15 Polyclonal specifically detects GDF-15 in Human, Primate samples. It is validated for Western Blot, ELISA Detection (Matched Antibody Pair).
Specifications
| GDF-15 | |
| Polyclonal | |
| Biotin | |
| Q99988 | |
| GDF15 | |
| Chinese hamster ovary cell line CHO-derived recombinant human GDF-15 (R&D Systems, Catalog # 957-GD) Ala197-Ile308 Accession # Q99988 | |
| 50 μg | |
| Primary | |
| Detects human GDF-15 in ELISAs and Western blots. In sandwich immunoassays, less than 0.1% cross-reactivity with recombinant human GDF-11, recombinant mouse (rm) GDF-5, rmGDF-6, rmGDF-7, and rmGDF-8 is observed. | |
| Human, Primate | |
| IgG |
| Western Blot, ELISA | |
| LYOPH | |
| Western Blot 0.1 ug/mL, ELISA Detection (Matched Antibody Pair) 0.1-0.4 ug/mL | |
| GDF15, growth differentiation factor 15, growth/differentiation factor 15, Macrophage inhibitory cytokine 1, MIC-1, MIC-1NSAID-activated gene 1 protein, MIC1Prostate differentiation factor, NAG-1, NAG-1NSAID-regulated gene 1 protein, NSAID (nonsteroidal inflammatory drug)-activated protein 1, PDF, PDFGDF-15, PLAB, PLABNRG-1, Placental bone morphogenetic protein, Placental TGF-beta, PTGF-beta, PTGFBPTGF-beta | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 9518 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction