Learn More
Human PRL/PTP4A Pan Specific Antibody, R&D Systems™
Mouse Monoclonal Antibody has been used in 4 publications
Brand: R&D Systems MAB32191-SP
This item is not returnable.
View return policy
Description
PRL/PTP4A Monoclonal specifically detects PRL/PTP4A in Human samples. It is validated for Immunohistochemistry.
Specifications
| PRL/PTP4A | |
| Monoclonal | |
| LYOPH | |
| Immunohistochemistry 8-25 ug/mL | |
| O75365 | |
| E. coli-derived recombinant human PRL-3 Ala2-Met173 Accession # O75365 | |
| 25 μg | |
| Primary | |
| Detects human PRL-2 and human PRL-3 with equal affinities in direct ELISAs. In direct ELISAs, this antibody shows 10% cross-reactivity with recombinant human PRL-1. | |
| Human | |
| Purified |
| Immunohistochemistry | |
| 334407 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 7803 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2b |