Learn More
Human PRL/PTP4A Pan Specific Antibody, R&D Systems™
Mouse Monoclonal Antibody has been used in 4 publications
Brand:  R&D Systems MAB32191
 View more versions of this product
Product Code. 18171171
  • € 377.00 / 100µg
Description
PRL/PTP4A Monoclonal specifically detects PRL/PTP4A in Human samples. It is validated for Immunohistochemistry.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
PRL/PTP4A
Monoclonal
LYOPH
Immunohistochemistry 8-25 ug/mL
O75365
E. coli-derived recombinant human PRL-3 Ala2-Met173 Accession # O75365
100 μg
Primary
Detects human PRL-2 and human PRL-3 with equal affinities in direct ELISAs. In direct ELISAs, this antibody shows 10% cross-reactivity with recombinant human PRL-1.
Human
Purified
Immunohistochemistry
334407
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Mouse
Protein A or G purified from hybridoma culture supernatant
RUO
7803
Reconstitute at 0.5 mg/mL in sterile PBS.
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
IgG2b