missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Lysine (K)-specific Demethylase 6B/KDM6B Antibody, R&D Systems™

Goat Polyclonal Antibody
Brand: R&D Systems AF7300-SP
This item is not returnable.
View return policy
Description
Lysine (K)-specific Demethylase 6B/KDM6B/JMJD3 Polyclonal specifically detects Lysine (K)-specific Demethylase 6B/KDM6B/JMJD3 in Human samples. It is validated for Immunocytochemistry.
Specifications
| Lysine (K)-specific Demethylase 6B/KDM6B/JMJD3 | |
| Polyclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| EC 1.14.11, EC 1.14.11.-, JmjC domain-containing protein 3, JMJD3, JMJD3lysine-specific demethylase 6B, jumonji domain containing 3, histone lysine demethylase, Jumonji domain-containing protein 3, KDM6B, KIAA0346jumonji domain containing 3, lysine (K)-specific demethylase 6B, Lysine (K)specific Demethylase 6B, Lysine demethylase 6B | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 23135 | |
| Sterile PBS to a final concentration of 0.2 mg/mL. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| Immunocytochemistry | |
| LYOPH | |
| Immunocytochemistry 5-15 ug/mL | |
| NP_001073893 | |
| KDM6B | |
| E. coli-derived recombinant human Lysine (K)-specific Demethylase 6B/KDM6B (isoform 1) Pro1530-Arg1682 Accession # NP_001073893 | |
| 25 μg | |
| Primary | |
| Detects human Lysine (K)-specific Demethylase 6B/KDM6B in direct ELISAs. | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction