missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Pro-Cathepsin B Biotinylated Antibody, R&D Systems™
Mouse Monoclonal Antibody
Brand: R&D Systems BAM21761
This item is not returnable.
View return policy
Description
Cathepsin B Monoclonal specifically detects Cathepsin B in Human samples. It is validated for ELISA Detection (Matched Antibody Pair).
Specifications
| Cathepsin B | |
| Monoclonal | |
| LYOPH | |
| ELISA Detection (Matched Antibody Pair) 0.5-2.0 ug/mL | |
| P07858 | |
| CTSB | |
| Mouse myeloma cell line NS0-derived recombinant human Cathepsin B Arg18-Ile339 Accession # P07858 | |
| 250 μg | |
| Primary | |
| Detects human Pro-Cathepsin B in sandwich ELISAs. | |
| Human | |
| Purified |
| ELISA | |
| 155709 | |
| Biotin | |
| Lyophilized from a 0.2 μm filtered solution in PBS with BSA as a carrier protein. with No Preservative | |
| APP secretase, APPS, cathepsin B, Cathepsin B1, CPSBamyloid precursor protein secretase, CTSB, cysteine protease, EC 3.4.22, EC 3.4.22.1 | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 1508 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction