Learn More
R&D Systems™ Human Pro-Collagen I alpha 1 DuoSet ELISA
Product Code. 30021625
Click to view available options
Quantity:
1 Kit (for 5 Plates)
Unit Size:
1 set
Product Code. 30021625
Brand: R&D Systems™ DY622005
Human Pro-Collagen I alpha 1 DuoSet ELISA from the most referenced ELISA manufacturer. Highly validated for accurate quantitation and long-term reproducibility.
The Human Pro-Collagen I alpha 1 DuoSet ELISA is a Solid Phase Sandwich ELISA that quantifies human Collagen I alpha 1.
TRUSTED_SUSTAINABILITY
Specifications
Quantity 1 Kit (for 5 Plates)
Species Human
Conjugate Biotin
Assay Range 31.20 to 2000 pg/mL
For Use With (Application) ELISA
Gene ID (Entrez) 1277
Synonym Alpha-1 type I collagen, COL1A1, COL-IA1, Collagen 1, Collagen 1 alpha 1, collagen alpha 1 chain type I, collagen alpha-1(I) chain, collagen of skin, tendon and bone, alpha-1 chain, Collagen Type I Alpha 1 Chain, collagen, type I, alpha 1, Collagen1, Collagen-1, EDSARTH1, OI4, pro-alpha-1 collagen type 1, Type I Procollagen Alpha 1 Chain
Content And Storage Store the unopened product at 2°C to 8°C. Do not use past expiration date.