missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Human Pro-MMP-1 Quantikine ELISA Kit

Product Code. 18722374
Click to view available options
Quantity:
1 Pk. (6 Plates)
1 Plate
Sufficient For:
1 x 96-well Plate
6 x 96-well Plates
Unit Size:
1 set
This item is not returnable. View return policy

Product Code. 18722374

Brand: R&D Systems™ DMP100

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Human Pro-MMP-1 Quantikine ELISA Kit from the most referenced ELISA manufacturer. Highly validated for accurate quantitation and long-term reproducibility.

The Human Pro-MMP-1 Quantikine ELISA Kit is a Solid Phase Sandwich ELISA that quantifies human MMP-1 in cell culture supernates (100 ul), serum (100 ul), heparin plasma (100 ul).
TRUSTED_SUSTAINABILITY

Specifications

Assay Range 0.16 to 10 ng/mL
Assay Sensitivity 0.095 ng/mL
Conjugate HRP
Cross Reactivity Cross-species reactivity not tested. <0.5% cross-reactivity observed with available related molecules.
Product Type Solid Phase Sandwich ELISA
Sample Type Cell Culture Supernatant, Plasma (Heparin), Serum
Specificity Natural and Recombinant Human Pro-MMP-1
Sufficient For 1 x 96-well Plate
Target Species Human
For Use With (Application) ELISA
Gene ID (Entrez) 4312
Gene Symbol MMP1
Quantity 1 Plate
Sample Volume Cell Culture Supernates (100uL),Serum (100uL),Heparin Plasma (100uL)
Storage Requirements Store the unopened product at 2°C to 8°C. Do not use past expiration date.
Synonym CLGmatrix metalloprotease 1, CLGN, EC 3.4.24, EC 3.4.24.7, Fibroblast collagenase, interstitial collagenase, matrix metallopeptidase 1 (interstitial collagenase), matrix metalloproteinase 1 (interstitial collagenase), Matrix metalloproteinase-1, MMP1, MMP-1
Target MMP-1
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.