missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Pro-Relaxin-1 Antibody, R&D Systems™

Goat Polyclonal Antibody
Brand: R&D Systems AF3257
This item is not returnable.
View return policy
Description
Relaxin-1 Polyclonal specifically detects Relaxin-1 in Human samples. It is validated for Western Blot.
Specifications
| Relaxin-1 | |
| Polyclonal | |
| Unconjugated | |
| P04808 | |
| RLN1 | |
| E. coli-derived recombinant human Relaxin-1 Lys26-Cys185 Accession # P04808 | |
| 100 μg | |
| Primary | |
| Detects human Relaxin-1 in direct ELISAs and Western blots. In direct ELISAs, less than 5% cross-reactivity with recombinant human (rh) Relaxin-2 and rhRelaxin-3 is observed. | |
| Human | |
| IgG |
| Western Blot | |
| LYOPH | |
| Western Blot 1 ug/mL | |
| bA12D24.3.1, bA12D24.3.2, H1, preprorelaxin H1, prorelaxin H1, relaxin 1, relaxin 1 (H1), relaxin H1, Relaxin1, RLN1, RLXH1 | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 6013 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction