missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Procalcitonin Antibody, R&D Systems™

Sheep Polyclonal Antibody
Brand: R&D Systems AF8350
This item is not returnable.
View return policy
Description
Procalcitonin Polyclonal specifically detects Procalcitonin in Human samples. It is validated for Western Blot.
Specifications
| Procalcitonin | |
| Polyclonal | |
| Western Blot 1 ug/mL | |
| P01258 | |
| CALCA | |
| E. coli-derived recombinant human Procalcitonin Ala26-Gly117 (Asn57Asp) Accession # P01258 | |
| 100 μg | |
| Primary | |
| Detects human Procalcitonin in direct ELISAs and Western blots. | |
| Human | |
| IgG |
| Western Blot | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| CALC1, CALCA, Calcitonin 1, calcitonin gene-related peptide 1, CGRP, Katacalcin | |
| Sheep | |
| Affinity Purified | |
| RUO | |
| 796 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction