missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Human Procalcitonin DuoSet ELISA
Description
The Human Procalcitonin DuoSet ELISA is a Solid Phase Sandwich ELISA that quantifies human Procalcitonin.
Specifications
Specifications
| Assay Range | 31.20 to 2,000pg/mL |
| Conjugate | Biotin |
| Immunoassay Kit Format | 96-well Strip Plate |
| Product Type | Solid Phase Sandwich ELISA |
| Sufficient For | Five 96-well plates |
| For Use With (Application) | ELISA |
| Storage Requirements | Store the unopened product at 2° to 8°C. Do not use past expiration date. |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?