missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Proprotein Convertase 2/PCSK2 Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems MAB6018-SP
This item is not returnable.
View return policy
Description
Proprotein Convertase 2/PCSK2 Monoclonal specifically detects Proprotein Convertase 2/PCSK2 in Human samples. It is validated for Western Blot, Immunoprecipitation, Immunocytochemistry.
Specifications
| Proprotein Convertase 2/PCSK2 | |
| Monoclonal | |
| LYOPH | |
| Western Blot 2 ug/mL, Immunoprecipitation 25 ug/mL, Immunocytochemistry 8-25 ug/mL | |
| P16519 | |
| PCSK2 | |
| Chinese hamster ovary cell line CHO-derived recombinant human Proprotein Convertase 2/PCSK2 Gly110-Asn638 Accession # P16519 | |
| 25 μg | |
| Primary | |
| Detects human Proprotein Convertase 2/PCSK2 in direct ELISAs. In direct ELISAs and Western blots, no cross-reactivity with recombinant human (rh) Proprotein Convertase 1, 7, 9, or rhFurin is observed. | |
| Human | |
| Purified |
| Western Blot, Immunoprecipitation, Immunocytochemistry | |
| 694009 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| EC 3.4.21, EC 3.4.21.94, KEX2-like endoprotease 2, NEC 2, NEC2, NEC2SPC2, neuroendocrine convertase 2, PC2, PC2NEC-2, PCSK2, Prohormone convertase 2, Proprotein convertase 2, proprotein convertase subtilisin/kexin type 2, SPC2 | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 5126 | |
| Sterile PBS to a final concentration of 0.5 mg/mL. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction