missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Protocadherin-1 Alexa Fluor 594-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems FAB5899T-100UG
This item is not returnable.
View return policy
Description
Protocadherin-1 Monoclonal specifically detects Protocadherin-1 in Human samples. It is validated for Flow Cytometry.
Specifications
| Protocadherin-1 | |
| Monoclonal | |
| 0.2 mg/mL | |
| Flow Cytometry 0.25-1 ug/10^6 cells | |
| Q08174 | |
| PCDH1 | |
| E. coli-derived recombinant human Protocadherin-1 Thr58-Asn162 Accession # Q08174 | |
| RUO | |
| 5097 | |
| Human | |
| IgG1 |
| Flow Cytometry | |
| 648127 | |
| Alexa Fluor 594 | |
| Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide | |
| cadherin-like 1, Cadherin-like protein 1, FLJ53887, MGC45991, PC42, PC42PCDH42, PCDH1, PCDH42, protocadherin 1, protocadherin 1 (cadherin-like 1), protocadherin 42, protocadherin-1, Protocadherin1, protocadherin-42 | |
| Mouse | |
| 100 μg | |
| Primary | |
| Detects human Protocadherin-1 in direct ELISAs. | |
| Store the unopened product at 2 - 8 degreesC. Do not use past expiration date. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction