Learn More
Human Protocadherin-15 Antibody, R&D Systems™
Sheep Polyclonal Antibody
Brand: R&D Systems AF6729
This item is not returnable.
View return policy
Description
Protocadherin-15 Polyclonal specifically detects Protocadherin-15 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry.
Specifications
| Protocadherin-15 | |
| Polyclonal | |
| Unconjugated | |
| Q96QU1 | |
| PCDH15 | |
| Chinese hamster ovary cell line CHO-derived recombinant human Protocadherin-15 Gln27-Ala1376 Accession # Q96QU1 | |
| 100 μg | |
| Primary | |
| Detects human Protocadherin-15 in direct ELISAs. | |
| Human | |
| IgG |
| Western Blot, Immunohistochemistry, Immunocytochemistry | |
| LYOPH | |
| Western Blot 2 ug/mL, Immunohistochemistry 5-15 ug/mL, Immunocytochemistry 5-15 ug/mL | |
| autosomal recessive 23, cadherin-related family member 15, CDHR15, DFNB23, DKFZp667A1711, PCDH15, protocadherin-15, Protocadherin15, protocadherin-related 15, USH1F | |
| Sheep | |
| Affinity Purified | |
| RUO | |
| 65217 | |
| Sterile PBS to a final concentration of 0.2 mg/mL. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |