missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human PRUNE (aa 316-382) Control Fragment Recombinant Protein
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
Description
Highest antigen sequence indentity to the following orthologs: Mouse (76%), Rat (76%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-55842 (PA5-55842. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.
Phosphodiesterase (PDE) that has higher activity toward cAMP than cGMP, as substrate. It plays a role in cell proliferation, is able to induce cell motility and acts as a negative regulator of NME1.
Specifications
Specifications
| Accession Number | Q86TP1 |
| Concentration | ≥5.0 mg/mL |
| For Use With (Application) | Blocking Assay, Control |
| Formulation | 1 M urea, PBS with no preservative; pH 7.4 |
| Gene ID (Entrez) | 58497 |
| Name | Human PRUNE (aa 316-382) Control Fragment |
| Quantity | 100 μL |
| Regulatory Status | RUO |
| Gene Alias | 9230112O05Rik; C130058A12; DRES17; DRES-17; Drosophila-related expressed sequence 17; Exopolyphosphatase PRUNE1; hPrune; H-PRUNE; HTcD37; protein prune homolog; Protein prune homolog 1; Prune; prune exopolyphosphatase; prune homolog; prune homolog (Drosophila); PRUNE1; PRUNEM1; Prune-M1 |
| Common Name | PRUNE |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction