Learn More
R&D Systems™ Human PSCA (NP_005663.2) VersaClone cDNA
Specifications
Specifications
| Gene | PSCA |
| Accession Number | NP_005663.2 |
| Gene ID (Entrez) | 8000 |
| Gene Alias | PRO232, prostate stem cell antigen |
| Content And Storage | Store the unopened product at -20°C to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date. |
| Species | Human |
| Quantity | 10 μg |