Learn More
R&D Systems™ Human PSG-6 Recombinant Protein, Carrier-Free
Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility.
Brand: R&D Systems™ 9318-P6-050
Description
The Recombinant Human PSG6 Protein is derived from HEK293. The Recombinant Human PSG6 Protein has been validated for the following applications: Bioactivity.Order Info
Ships at ambient temperature.
Specifications
| Q00889-2 | |
| 3730 | |
| 50μg | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution. | |
| <0.10 EU per 1μg of the protein by the LAL method | |
| Recombinant protein | |
| Yes |
| Lyophilized from a 0.2μm filtered solution in PBS and NaCl with Trehalose | |
| 45 kDa | |
| Human embryonic kidney cell,HEK293-derived human PSG6 protein Gln35-His424,with a C-terminal 6-His tag | |
| RUO | |
| 0.9 to 4.5μg/mL | |
| Unconjugated |
For Research Use Only.