Learn More
R&D Systems™ Human PSG-6 Recombinant Protein, Carrier-Free
Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility.
Brand:  R&D Systems™ 9318-P6-050
Encompass
Product Code. 15770424
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Description
Description
The Recombinant Human PSG6 Protein is derived from HEK293. The Recombinant Human PSG6 Protein has been validated for the following applications: Bioactivity.
Order Info
Ships at ambient temperature.
Specifications
Specifications
Q00889-2
3730
50μg
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
<0.10 EU per 1μg of the protein by the LAL method
Recombinant protein
Yes
Lyophilized from a 0.2μm filtered solution in PBS and NaCl with Trehalose
45 kDa
Human embryonic kidney cell,HEK293-derived human PSG6 protein Gln35-His424,with a C-terminal 6-His tag
RUO
0.9 to 4.5μg/mL