missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human PSGR Antibody, R&D Systems™
Mouse Monoclonal Antibody
€ 209.00 - € 440.00
Specifications
| Antigen | PSGR |
|---|---|
| Clone | 1033009 |
| Dilution | Immunohistochemistry 5-25 ug/mL |
| Applications | Immunohistochemistry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18119160
|
R&D Systems
MAB10669-SP |
25 μg |
€ 209.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18103521
|
R&D Systems
MAB10669-100 |
100 μg |
€ 440.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
PSGR Monoclonal antibody specifically detects PSGR in Human samples. It is validated for ImmunohistochemistrySpecifications
| PSGR | |
| Immunohistochemistry 5-25 ug/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied either lyophilized or as a 0.2 μm filtered solution in PBS. | |
| HPRAJ, Olfactory Receptor OR11-16, olfactory receptor, family 51, subfamily E, member 2, olfactory receptor, family 51, subfamily E, member 3 pseudogene, olfactory receptor, family 52, subfamily A, member 2, OR51E2, OR51E3P, OR52A2, prostate specific G-protein coupled receptor, Prostate-specific G-protein coupled receptor, PSGRolfactory receptor 51E2 | |
| Human PSGR synthetic peptide, Accession # Q9H255 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. |
| 1033009 | |
| Immunohistochemistry | |
| Unconjugated | |
| Mouse | |
| Human | |
| Q9H255 | |
| 81285 | |
| IgG1 | |
| Protein A or G purified from hybridoma culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20°C to -70°C as supplied. 1 month, 2°C to 8°C under sterile conditions after reconstitution. 6 months, -20°C to -70°C under sterile conditions after reconstitution. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title