missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human PTH Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems MAB7665
This item is not returnable.
View return policy
Description
PTH Monoclonal specifically detects PTH in Human samples. It is validated for Western Blot, Immunohistochemistry.
Specifications
| PTH | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. with No Preservative | |
| Parathormone, Parathyrin, parathyroid hormone, parathyroid hormone 1, PTH1 | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 5741 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2b |
| Western Blot, Immunohistochemistry | |
| 918462 | |
| Western Blot 2 ug/mL, Immunohistochemistry 8-25 ug/mL | |
| P01270 | |
| PTH | |
| E. coli-derived recombinant human PTH Ser32-Gln115 Accession # P01270 | |
| 100 μg | |
| Primary | |
| Detects human PTH in direct ELISAs. | |
| Human | |
| Supernatant |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction