Learn More
Human PTS (PTPS) knockout HeLa cell lysate, 1 Kit
Product Code. 32141572 Shop All Abcam Products
Product Code. 32141572 Supplier Abcam Supplier No. ab2633221Kit
 Restricted Item
Estimated Shipment: 10-11-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Human PTS (PTPS) knockout HeLa cell/tissue lysate.
  • Species: Human
  • Tissue: PTS (PTPS) knockout HeLa