missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Pygopus-2 Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems MAB3616-SP
This item is not returnable.
View return policy
Description
Pygopus-2 Monoclonal specifically detects Pygopus-2 in Human samples. It is validated for Western Blot, Simple Western, Immunohistochemistry, Immunocytochemistry.
Specifications
| Pygopus-2 | |
| Monoclonal | |
| LYOPH | |
| Western Blot 0.5 ug/mL, Simple Western 5 ug/mL, Immunohistochemistry 3-25 ug/mL, Immunocytochemistry 8-25 ug/mL | |
| NP_612157 | |
| PYGO2 | |
| E. coli-derived recombinant human Pygopus-2 Met160-Glu336 Accession # NP_612157 | |
| 25 μg | |
| Primary | |
| Detects human Pygopus-2 in direct ELISAs and Western blots. In direct ELISAs, no cross-reactivity with recombinant human Pygopus-1 or recombinant mouse Pygopus-1 is observed. | |
| Human | |
| Purified |
| Western Blot, Immunohistochemistry, Immunocytochemistry | |
| 539730 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| 1190004M21Rik, FLJ33226, PYGO2, pygopus 2, pygopus homolog 2, pygopus homolog 2 (Drosophila), Pygopus2 | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 90780 | |
| Sterile PBS to a final concentration of 0.5 mg/mL. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction