Learn More
ACROBiosystems Human R-Spondin 3 / RSPO3 (22-146) Protein, His Tag
Description
R-Spondin 3 (RSPO3) is also called cysteine-rich and single thrombospondin domain containing-1 (CRISTIN1), Protein with TSP type-1 repeat (PWTSR), is a member of the R-spondin protein family. R-spondins (RSPO) are a recently discovered secretory protein family with four members in human and mouse. Although all four RSPO proteins activate the canonical Wnt pathway, RSPO2 and RSPO3 are more potent than RSPO1, whereas RSPO4 is relatively inactive. RSPO-3 is expressed ubiquitously and expressed at higher level in placenta, small intestine, fetal thymus and lymph node. RSPO3 is the activator of the beta-catenin signaling cascade, leading to TCF-dependent gene activation. RSPO3 acts both in the canonical Wnt/beta-catenin-dependent pathway and in non-canonical Wnt signaling pathway, probably by acting as an inhibitor of ZNRF3, an important regulator of the Wnt signaling pathway. RSPO3 also acts as a ligand for frizzled FZD8 and LRP6 and may negatively regulate the TGF-beta pathway.
Specifications
Specifications
| Accession Number | AAH22367.1 |
| Molecular Weight (g/mol) | 15.9 kDa |
| Quantity | 50 μg |
| Source | Human R-Spondin 3 (22-146), His Tag (RS6-H5222) is expressed from human 293 cells (HEK293). It contains AA Gln 22 - Val 146 (Accession # AAH22367.1). |
| Immunogen | Gln 22 - Val 146 |
| Storage Requirements | -20°C |
| Gene Alias | RSPO3,CRISTIN1,PWTSR,THSD2 |
| Common Name | R-Spondin 3 |
| Conjugate | Unconjugated |
| Species | Human |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.