missing translation for 'onlineSavingsMsg'
Learn More

ACROBiosystems Human R-Spondin 3 / RSPO3 (22-146) Protein, His Tag

Product Code. 15847627
Change view
Click to view available options
Quantity:
1 mg
50 μg
Unit Size:
1mg
50µg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15847627 1 mg 1mg
15857627 50 μg 50µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15847627 Supplier ACROBiosystems Supplier No. RS6H52221MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

R-Spondin 3 (RSPO3) is also called cysteine-rich and single thrombospondin domain containing-1 (CRISTIN1), Protein with TSP type-1 repeat (PWTSR), is a member of the R-spondin protein family. R-spondins (RSPO) are a recently discovered secretory protein family with four members in human and mouse. Although all four RSPO proteins activate the canonical Wnt pathway, RSPO2 and RSPO3 are more potent than RSPO1, whereas RSPO4 is relatively inactive. RSPO-3 is expressed ubiquitously and expressed at higher level in placenta, small intestine, fetal thymus and lymph node. RSPO3 is the activator of the beta-catenin signaling cascade, leading to TCF-dependent gene activation. RSPO3 acts both in the canonical Wnt/beta-catenin-dependent pathway and in non-canonical Wnt signaling pathway, probably by acting as an inhibitor of ZNRF3, an important regulator of the Wnt signaling pathway. RSPO3 also acts as a ligand for frizzled FZD8 and LRP6 and may negatively regulate the TGF-beta pathway.

This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 15.9 kDa. The protein migrates as 16 kDa and 19-21 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.

Specifications

Accession Number AAH22367.1
Molecular Weight (g/mol) 15.9 kDa
Quantity 1 mg
Source Human R-Spondin 3 (22-146), His Tag (RS6-H5222) is expressed from human 293 cells (HEK293). It contains AA Gln 22 - Val 146 (Accession # AAH22367.1).
Immunogen Gln 22 - Val 146
Storage Requirements -20°C
Gene Alias RSPO3,CRISTIN1,PWTSR,THSD2
Common Name R-Spondin 3
Conjugate Unconjugated
Species Human
Protein Tag His Tag
Buffer PBS, pH7.4
Endotoxin Level Less than 1.0 EU / μg by the LAL method.
Expression System HEK293
Form Powder
Protein Subtype Bio-Markers and CD Antigens
Purity or Quality Grade >90% as determined by SDS-PAGE
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.