missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human RAB25 Antibody, R&D Systems™

Goat Polyclonal Antibody
Brand: R&D Systems AF6605-SP
This item is not returnable.
View return policy
Description
Rab25 Polyclonal specifically detects Rab25 in Human samples. It is validated for Western Blot.
Specifications
| Rab25 | |
| Polyclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| CATX8, CATX-8RAB11C, RAB11C, RAB25, member RAS oncogene family, ras-related protein Rab-25 | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 57111 | |
| Sterile PBS to a final concentration of 0.2 mg/mL. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| Western Blot | |
| LYOPH | |
| Western Blot 2 ug/mL | |
| P57735 | |
| RAB25 | |
| E. coli-derived recombinant human RAB25 Met1-Ala195 Accession # P57735 | |
| 25 μg | |
| Primary | |
| Detects human RAB25 in Western blots. | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction