missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human/Rat beta-1,3-N-acetylglucosaminyl-transferase 2/B3GNT2 Antibody, R&D Systems™
Goat Polyclonal Antibody
Brand: R&D Systems AF3960
This item is not returnable.
View return policy
Description
Beta-1,3-N-Acetylglucosaminyltransferase 2/B3GNT2 Polyclonal specifically detects Beta-1,3-N-Acetylglucosaminyltransferase 2/B3GNT2 in Human, Rat samples. It is validated for Western Blot.
Specifications
| Beta-1,3-N-Acetylglucosaminyltransferase 2/B3GNT2 | |
| Polyclonal | |
| Unconjugated | |
| Q9NY97 | |
| B3GNT2 | |
| Mouse myeloma cell line NS0-derived recombinant human beta-1,3-N-acetylglucosaminyltransferase 2/B3GNT2 Lys29-Cys397 Accession # Q9NY97 | |
| 100 μg | |
| Primary | |
| Detects human and rat beta-1,3-N-acetylglucosaminyltransferase 2/B3GNT2 in Western blots. | |
| Human, Rat | |
| IgG |
| Western Blot | |
| LYOPH | |
| Western Blot 1 ug/mL | |
| B3Galt6, b3Gal-T7, B3GALT7, B3GNT, B3GNT1, B3GN-T1, B3GN-T2, B3GNT-2, Beta-1,3-galactosyltransferase 7, Beta-1,3-GalTase 7, beta-1,3-Gn-T1, Beta-1,3-Gn-T2, Beta-1,3-N-acetylglucosaminyltransferase 1, Beta-1,3-N-acetylglucosaminyltransferase 2, beta-1,3-N-acetylglucosaminyltransferase bGnT-1, beta-1,3-N-acetylglucosaminyltransferase bGnT-2, Beta3GalT7, Beta3Gal-T7, BETA3GNT, Beta3gnt1, Beta3Gn-T1, beta3Gn-T2, Beta-3-Gx-T7, BGnT-1, BGnT-2, BGNT2, EC 2.4.1.-, UDP-Gal:beta-GlcNAc beta-1,3-galactosyltransferase 7, UDP-galactose:beta-N-acetylglucosamine beta-1,3-galactosyltransferase 7, UDP-GlcNAc:betaGal beta-1,3-N-acetylglucosaminyltransferase 1, UDP-GlcNAc:betaGal beta-1,3-N-acetylglucosaminyltransferase 2 | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 10678 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction