missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human/Rat Neuroligin 1/NLGN1 Antibody, R&D Systems™

Sheep Polyclonal Antibody
Brand: R&D Systems AF4340-SP
This item is not returnable.
View return policy
Description
Neuroligin 1/NLGN1 Polyclonal specifically detects Neuroligin 1/NLGN1 in Human, Rat samples. It is validated for Western Blot, Immunohistochemistry.
Specifications
| Neuroligin 1/NLGN1 | |
| Polyclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| EC 3.1.1, EC 3.1.1.1, KIAA1070neuroligin-1, MGC45115, neuroligin 1, NL1, Nlg, NLGN1 | |
| Sheep | |
| Affinity Purified | |
| RUO | |
| 22871 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| Western Blot, Immunohistochemistry | |
| LYOPH | |
| Western Blot 0.1 ug/mL, Immunohistochemistry 5-15 ug/mL | |
| Q62765 | |
| NLGN1 | |
| Mouse myeloma cell line NS0-derived recombinant rat Neuroligin 1/NLGN1 Gln46-Leu696 Accession # Q62765 | |
| 25 μg | |
| Primary | |
| Detects recombinant rat Neuroligin 1/NLGN1 in direct ELISAs. Detects human and rat Neuroligin 1/NLGN1 in Western blots. | |
| Human, Rat | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction