missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human/Rat Neuroligin 1/NLGN1 Antibody, R&D Systems™

Sheep Polyclonal Antibody
Brand: R&D Systems AF4340
This item is not returnable.
View return policy
Description
Neuroligin 1/NLGN1 Polyclonal specifically detects Neuroligin 1/NLGN1 in Human, Rat samples. It is validated for Western Blot, Immunohistochemistry.
Specifications
| Neuroligin 1/NLGN1 | |
| Polyclonal | |
| Unconjugated | |
| Q62765 | |
| NLGN1 | |
| Mouse myeloma cell line NS0-derived recombinant rat Neuroligin 1/NLGN1 Gln46-Leu696 Accession # Q62765 | |
| 100 μg | |
| Primary | |
| Detects recombinant rat Neuroligin 1/NLGN1 in direct ELISAs. Detects human and rat Neuroligin 1/NLGN1 in Western blots. | |
| Human, Rat | |
| IgG |
| Western Blot, Immunohistochemistry | |
| LYOPH | |
| Western Blot 0.1 ug/mL, Immunohistochemistry 5-15 ug/mL | |
| EC 3.1.1, EC 3.1.1.1, KIAA1070neuroligin-1, MGC45115, neuroligin 1, NL1, Nlg, NLGN1 | |
| Sheep | |
| Affinity Purified | |
| RUO | |
| 22871 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction