missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human/Rat OV-6 Alexa Fluor 488-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems FAB2020G-100UG
This item is not returnable.
View return policy
Description
OV-6 Monoclonal specifically detects OV-6 in Human, Rat samples. It is validated for Flow Cytometry.
Specifications
| OV-6 | |
| Monoclonal | |
| 0.2 mg/mL | |
| Flow Cytometry 0.25-1 ug/10^6 cells | |
| Mouse | |
| 100 μg | |
| Primary | |
| Human, Rat | |
| IgG1 |
| Flow Cytometry | |
| OV-6 | |
| Alexa Fluor 488 | |
| Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide | |
| Carcinogen-treated rat nodular hepatocytes | |
| RUO | |
| Detects human and rat OV-6 antigen. | |
| Store the unopened product at 2 - 8 degreesC. Do not use past expiration date. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction