Learn More
Human/Rat Tollip Antibody, R&D Systems™
GREENER_CHOICE
Mouse Monoclonal Antibody
Brand:  R&D Systems MAB4678-SP
Product Code. 18761269
  • € 209.00 / 25µg
Description
Tollip Monoclonal specifically detects Tollip in Human, Rat samples. It is validated for Western Blot.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Tollip
Monoclonal
LYOPH
Western Blot 1 ug/mL
Q9H0E2
TOLLIP
E. coli-derived recombinant human Tollip Met1-Pro274 Accession # Q9H0E2
25 μg
Primary
Detects human and rat Tollip in Western blots.
Human, Rat
Purified
Western Blot
503315
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
adapter protein, FLJ33531, IL-1RAcPIP, toll interacting protein, toll-interacting protein
Mouse
Protein A or G purified from hybridoma culture supernatant
RUO
54472
Reconstitute at 0.5 mg/mL in sterile PBS.
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
IgG2a