missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human RBBP6 (aa 1295-1383) Control Fragment Recombinant Protein
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
Description
Highest antigen sequence indentity to the following orthologs: Mouse (82%), Rat (82%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-111180 (PA5-111180. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.
This gene encodes a cilia-associated protein belonging to the kinesin family. This protein plays a role in the sonic hedgehog (SHH) signaling pathway through the regulation of GLI transcription factors. It functions as a negative regulator of the SHH pathway by preventing inappropriate activation of GLI2 in the absence of ligand, and as a positive regulator by preventing the processing of GLI3 into its repressor form. Mutations in this gene have been associated with various ciliopathies.
Specifications
Specifications
| Accession Number | Q7Z6E9 |
| Concentration | ≥5.0 mg/mL |
| For Use With (Application) | Blocking Assay, Control |
| Formulation | 1 M urea, PBS with no preservative; pH 7.4 |
| Gene ID (Entrez) | 5930 |
| Name | Human RBBP6 (aa 1295-1383) Control Fragment |
| Quantity | 100 μL |
| Regulatory Status | RUO |
| Gene Alias | 4933422O15Rik; AI316869; BB233631; C030034J04Rik; C77662; E3 ubiquitin-protein ligase RBBP6; im:7158869; MY038; my038-like; P2PR; P2P-R; p53-associated cellular protein of testis; PACT; Proliferation potential-related protein; protein P2P-R; RB binding protein 6, ubiquitin ligase; RB-binding Q-protein 1; Rbbp6; rbbp6l; RBQ1; RBQ-1; retinoblastoma binding protein 6; retinoblastoma-binding protein 6; retinoblastoma-binding protein 6, isoform 3; retinoblastoma-binding Q protein 1; RING-type E3 ubiquitin transferase RBBP6; SNAMA |
| Common Name | RBBP6 |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction