missing translation for 'onlineSavingsMsg'
Learn More

STEMCELL Technologies Human Recombinant CNTF, ACF

Product Code. 17179741
Encompass_Preferred
Click to view available options
Quantity:
100 μg
1000 μg
20 μg
Unit Size:
1000µg
100µg
20µg
This item is not returnable. View return policy

Product Code. 17179741

Brand: STEMCELL Technologies 78170

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Ciliary neurotrophic factor, animal component-free

  • Ciliary neurotrophic factor (CNTF) is a neurotrophic factor that belongs to the four-helix bundle cytokine family and is structurally related to interleukin 6 (IL-6), interleukin 11 (IL-11), leukemia inhibitory factor (LIF), and oncostatin M (OSM)
  • CNTF binds to its receptor CNFTRα and induces formation of a heterodimer of the signal transducing IL-6 receptor gp130 and LIF receptor (LIFR)-β, which triggers JAK/STAT, ERK, and PI3K signaling cascades
  • CNTF plays an important role in neurogenesis and the differentiation of neural stem cells and has been suggested to possess a therapeutic role in treating neurological disorders
  • Additionally, CNTF is commonly used to differentiate human pluripotent stem cell (hPSC)-derived neural progenitor cells into astrocytes
  • This product is animal component-free

Specifications

Name Ciliary neurotrophic factor, HCNTF
Quantity 20 μg
Cell Type Neural Cells, PSC-Derived; Neural Stem and Progenitor Cells; Neurons
Purity or Quality Grade ≥95%
Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.