missing translation for 'onlineSavingsMsg'
Learn More

STEMCELL Technologies Human Recombinant DKK-1

Product Code. 17119841
Encompass_Preferred
Click to view available options
Quantity:
1000 μg
50 μg
Unit Size:
1000µg
50µg
This item is not returnable. View return policy

Product Code. 17119841

Brand: STEMCELL Technologies 78208.1

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Dickkopf-related protein 1

  • Dickkopf-related protein 1 (DKK-1) is a member of the Dickkopf family and is a secreted protein that inhibits the canonical WNT pathway by competitive binding to low-density lipoprotein receptors (LRP)-5 and 6 with high affinity, thereby decreasing β-catenin protein stability (Niehrs)
  • DKK-1 regulates embryonic development and contains two conserved cysteine-rich domains separated by a linker region and an N-terminal signal peptide
  • A family of human DKK-related genes composed of DKK-1, DKK-2, DKK-3, and DKK-4 have been characterized together with a unique DKK-3 related protein termed Soggy
  • DKK-1 has been shown to support the generation of myeloid-derived suppressor cells (MDSCs) and thus is a negative regulator of antitumor immune responses
  • DKK-1 from thrombocytes is an important regulator of leukocyte infiltration and induces Th2 cell polarization and potentiates

Specifications

Name Dickkopf-1, Dickkopf WNT signaling pathway inhibitor 1, hDkk-1, SK
Quantity 50 μg
Cell Type Endoderm, PSC-Derived; Myeloid Cells; Pluripotent Stem Cells
Purity or Quality Grade ≥95%
Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.