missing translation for 'onlineSavingsMsg'
Learn More
Learn More
STEMCELL Technologies Human Recombinant DKK-1
Click to view available options
Quantity:
1000 μg
50 μg
Unit Size:
1000µg
50µg
Description
- Dickkopf-related protein 1 (DKK-1) is a member of the Dickkopf family and is a secreted protein that inhibits the canonical WNT pathway by competitive binding to low-density lipoprotein receptors (LRP)-5 and 6 with high affinity, thereby decreasing β-catenin protein stability (Niehrs)
- DKK-1 regulates embryonic development and contains two conserved cysteine-rich domains separated by a linker region and an N-terminal signal peptide
- A family of human DKK-related genes composed of DKK-1, DKK-2, DKK-3, and DKK-4 have been characterized together with a unique DKK-3 related protein termed Soggy
- DKK-1 has been shown to support the generation of myeloid-derived suppressor cells (MDSCs) and thus is a negative regulator of antitumor immune responses
- DKK-1 from thrombocytes is an important regulator of leukocyte infiltration and induces Th2 cell polarization and potentiates
Specifications
Specifications
| Name | Dickkopf-1, Dickkopf WNT signaling pathway inhibitor 1, hDkk-1, SK |
| Quantity | 50 μg |
| Cell Type | Endoderm, PSC-Derived; Myeloid Cells; Pluripotent Stem Cells |
| Purity or Quality Grade | ≥95% |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction